missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, reagent grade, ≥98%, contains ∼2% ethanol and ∼10ppm BHT as inhibitor, Honeywell

Product Code. 15694310 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15694310 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694310 Supplier Honeywell Research Chemicals Supplier No. 3461441L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

reagent grade,≥98%, contains ∼2% ethanol and ∼10ppm BHT as inhibitor

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -116°C (literature)
CAS 60-29-7
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point −45°C
Packaging Glass Bottle
Molecular Formula C4H10O
Linear Formula C20H14O4
MDL Number MFCD00011646
UN Number UN1155
Beilstein 1696894
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Boiling Point 34°C to 35°C
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12
Percent Purity ≥98%
Grade Reagent
Assay BHT
Chemical Name or Material Diethyl ether, reagent grade, ≥98%, contains ∼2% ethanol and ∼10ppm BHT as inhibitor
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.