missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, puriss. p.a., ACS Reagent, Reag. Ph. Eur. reag. ISO, ≥99.8% (GC), Solstice

Product Code. 15653680
Change view
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
Pack of 4
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15653680 4 x 2.5 L Glass Bottle Pack of 4
15693680 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15653680 Supplier Honeywell-Riedel-de Haen Supplier No. 322034X2.5L
  • ONLINE EXCLUSIVE PRICE
    £407.00
    £351.27 / Pack of 4
    Save £55.73
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., contains BHT as inhibitor, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Appearance Liquid
Density 0.706g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Packaging Glass Bottle
Refractive Index n20/D 1.3530
Residue after Evaporation 0 to 0.001 %
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34.6°C
Formula Weight 74.12g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Diethyl Ether, Reag. Ph. Eur. reag. ISO
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.