Learn More
Diethyl Ether, for HPLC, Stabilised with Ethanol, Fisher Chemical™
Brand: Fisher Chemical D/2506/PB17
Code : 25
Additional Details : CAS Number : 60-29-7 Weight : 1.78250kg Transport : UN number : 1155 Chem class : 3 Pack group : I
Packaging | Plastic Coated Glass bottle |
---|---|
Quantity | 2.5L |
Chemical Identifiers
60-29-7 | |
74.12 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
3283 | |
ethoxyethane |
C4H10O | |
MFCD00011646 | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CHEBI:35702 | |
CCOCC |
Specifications
60-29-7 | |
MFCD00011646 | |
RTZKZFJDLAIYFH-UHFFFAOYSA-N | |
ethoxyethane | |
3283 | |
74.12g/mol | |
34.6°C | |
-116°C | |
Plastic Coated Glass bottle | |
587mbar at 20°C |
C4H10O | |
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether | |
CCOCC | |
74.12 | |
CHEBI:35702 | |
Liquid | |
Colorless | |
2.5L | |
2.55 | |
0.2448cp at 20°C |
Safety and Handling
- Diethyl ether
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.