missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diethyl ether, HPLC Grade, 99%, stab. with ethanol

Product Code. 11329057
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329057 1 L 1 L
11399047 1 L 1 L
15466275 4 x 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329057 Supplier Thermo Scientific Chemicals Supplier No. 038990.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents. | CAS: 60-29-7 | C4H10O | 74.12 g/mol

It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl ether
Melting Point °C to 116°C
CAS 60-29-7
Density 0.713 g/mL
Quantity 1 L
Flash Point -45°C (-49°F)
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula C4H10O
MDL Number MFCD00011646
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12
Grade HPLC
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.