missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%, Solstice

Product Code. 15653150
Change view
Click to view available options
Quantity:
100 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15653150 100 mL Glass Bottle 100mL
15663150 1 L Glass Bottle 1 L
15673150 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15653150 Supplier Honeywell-Chromasolv Supplier No. 309966100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, for HPLC, inhibitor-free,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
Absorbance max. 1.00 at 218 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 250 nm
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 100 mL
Flash Point −45°C
Packaging Glass Bottle
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Diethyl ether, CHROMASOLV™, for HPLC, inhibitor-free, ≥99.9%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.