missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ChromAR™ for HPLC, Macron Fine Chemicals™

Product Code. 15518734
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15518734 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518734 Supplier Macron Fine Chemicals Supplier No. 672725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For High Performance Liquid Chromatography

Specifications

CAS 60-29-7
Quantity 2.5 L
Molecular Formula C4H10O
MDL Number MFCD00011646
UN Number 1155
Synonym Ether, 2,2 '-Oxybisethane, Ethoxyethane, Ethyl Ether, 2,2 '-Oxydiethane
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.