missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Anhydrous, BAKER ANALYZED, J.T. Baker™

Product Code. 10362981 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10362981 5 L EcoTainer Metal Safety Can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10362981 Supplier JT Baker Supplier No. 8401.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade chemical stabilized with 7ppm BHT for laboratory and specialized industrial use.

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -116.3°C
CAS 60-29-7
Color Colorless
Density 0.71g/cm3 at 20°C
Quantity 5 L
Explosion Limit 1.7 - 48 Vol %
Flash Point −40°C
Ignition Point 170°C
Odor Sweetish
Packaging EcoTainer Metal Safety Can
Physical Form Fluid
Residue after Evaporation Max. 0.001%
Molecular Formula C4H10O
Linear Formula (C2H5)2O
MDL Number MFCD00011646
Viscosity 0.233 mPass at 20°C
Water Max. 0.01%, 12g/L
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
Boiling Point 35°C
Solubility Information 12g/L in water
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12g/mol
Percent Purity ≥99.5%
Grade BAKER ANALYZED
Chemical Name or Material Diethyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.