missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl ether, ACS Reagent, contains BHT as inhibitor, anhydrous, ≥99.0%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
250 mL
1 L
6 x 1 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -116°C (literature) |
CAS Max % | ≤100.0000% |
Density | 0.706 g/mL at 25 °C(lit.) |
Boiling Point | 34°C to 35°C |
Quantity | 6 x 1 L |
Flash Point | −45°C |
Packaging | Glass Bottle |
Physical Form | anhydrous |
Linear Formula | (CH3CH2)2O |
UN Number | UN1155 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Diethyl ether
- Danger
- Flammable liquid Category 1
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H302-Harmful if swallowed.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'