missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%, Solstice

Product Code. 15636680
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 4 L
Packaging:
Aluminum Bottle
Glass Bottle
Unit Size:
1 L
250mL
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15636680 4 x 4 L Glass Bottle Pack of 4
15626680 250 mL Glass Bottle 250mL
15616680 1 L Aluminum Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15636680 Supplier Honeywell-Riedel-de Haen Supplier No. 6738114X4L
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

ACS reagent, anhydrous, contains BHT as inhibitor,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C (literature)
CAS Max % ≤100.0000%
Density 0.706 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point −45°C
Packaging Glass Bottle
Physical Form anhydrous
Linear Formula (CH3CH2)2O
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C
Formula Weight 74.12
Percent Purity >99%
Grade ACS
Chemical Name or Material Diethyl ether, ACS reagent, anhydrous, contains BHT as inhibitor, ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.