missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, ACS, 98% min, stab. with 0.001% BHT and 3% ethanol

Product Code. 11347538
Change view
Click to view available options
Quantity:
500 g
6 x 500 g
Unit Size:
3000g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347538 6 x 500 g 3000g
11357538 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347538 Supplier Thermo Scientific Alfa Aesar Supplier No. 033224.62
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Widely used as a solvent, dissolves P, S, Br2 and I2 | CAS: 60-29-7 | C4H10O | 74.12 g/mol

Widely used as a solvent. Dissolves P, S, Br2 and I2. It is a widely used as a solvent in organic synthesis, and in the preparation of Grignard reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Color Colorless
Density 0.713 g/mL
Quantity 6 x 500 g
Flash Point -45°C (-49°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.3525
Assay Percent Range 98% min
Linear Formula (C2H5)2O
UN Number UN1155
Merck Index 14,3806
Boiling Point 34.6°C
Solubility Information Immiscible with water,miscible with alcohol.
Formula Weight 74.12
Percent Purity ≤98%
Grade ACS
Chemical Name or Material Diethyl ether, Stabilized with 0.001% BHT and 3% ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.