missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free, Thermo Scientific™

Catalog Number 11385307
Click to view available options
Quantity:
1L
4L
4 x 1L
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC
This item is not returnable. View return policy
60-29-7
C4H10O
74.12
MFCD00011646
RTZKZFJDLAIYFH-UHFFFAOYSA-N
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
3283
CHEBI:35702
ethoxyethane
CCOCC

Solvent For U.V. Spectrophotometric and General Use

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Assay Percent Range >99%
Appearance Colorless liquid
UN Number 1155
Formula Weight 74.12
Grade Sprectrophotometric
Density 0.713g/cm3 at 20°C
Boiling Point 34.6°C
Quantity 4L
Flash Point −45°C
Ignition Point 170°C (338°F)
Odor Sweet
Vapor Pressure 586.3 hPa (440mm Hg) at 20°C
Chemical Name or Material Diethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'