missing translation for 'onlineSavingsMsg'
Learn More

Solstice Diethyl Ether

Product Code. 15945430
Change view
Click to view available options
Quantity:
18 L
45 L
Unit Size:
18 L
45 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15945430 18 L 18 L
15955430 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15945430 Supplier Solstice Supplier No. 2400418L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Diethyl Ether, Puriss., meets analytical specification of BP, Ph. Eur., ≥99.5% (GC).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym Ether, Ethyl ether
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

pH Not applicable
Melting Point -116°C (1.013 hPa)
Color Colorless
CAS Max % ≤100.00%
Density 0.71 g/cm3 (20°C)
Quantity 18 L
Flash Point -45°C (1.013 hPa)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss.
Linear Formula (CH3CH2)2O
Vapor Pressure 716 hPa (25°C)
Viscosity 0.24 mPa.s (20°C)
UN Number UN1155
Beilstein 1696894
Boiling Point 34°C to 35°C (1.013 hPa)
Percent Purity ≥99.50%
Grade Pharmacopeia
Chemical Name or Material Diethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.