missing translation for 'onlineSavingsMsg'
Learn More

Diethyl dibromomalonate, 96%

Product Code. 11484493
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484493 100 g 100g
11394499 5 g 5g
11474493 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484493 Supplier Thermo Scientific Alfa Aesar Supplier No. A11508.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 631-22-1 | C7H10Br2O4 | 317.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-22-1
Molecular Formula C7H10Br2O4
Molecular Weight (g/mol) 317.96
MDL Number MFCD00015154
InChI Key PFZYFZRUPFUEOB-UHFFFAOYSA-N
PubChem CID 69427
IUPAC Name diethyl 2,2-dibromopropanedioate
SMILES CCOC(=O)C(Br)(Br)C(=O)OCC

Specifications

Density 1.68
Boiling Point 108°C to 110°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.485
Quantity 100 g
UN Number UN1760
Beilstein 1783879
Synonym diethyl dibromomalonate, diethyl 2,2-dibromomalonate, ethyl dibromomalonate, propanedioic acid, dibromo-, diethyl ester, malonic acid, dibromo-, diethyl ester, propanedioic acid, 2,2-dibromo-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibromopropanedioate, diethyldibromomalonate, acmc-1cuuu, diethyl2,2-dibromomalonate
Formula Weight 317.96
Percent Purity 97%
Chemical Name or Material Diethyl dibromomalonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.