missing translation for 'onlineSavingsMsg'
Learn More

Diethyl diallylmalonate, 97+%

Product Code. 11479693
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479693 50 g 50g
11469693 10 g 10g
11489693 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479693 Supplier Thermo Scientific Alfa Aesar Supplier No. A12460.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3195-24-2 | C13H20O4 | 240.299 g/mol

Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke?s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl diallylmalonate is the starting material for enantioselective synthesis of carbocyclic nucleoside analogues. It is also used as Low catalyst loading in ring-closing metathesis reaction. Cationic nickel (with monodentate phosphoramidites and Wilke′s azaphospholene as ligands) catalyzed cycloisomerisation of diethyl diallylmalonate has been reported.

Solubility
Sparingly Soluble in water. (0.72 g/L) (25°C)

Notes
Store under cool dry place. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3195-24-2
Molecular Formula C13H20O4
Molecular Weight (g/mol) 240.299
MDL Number MFCD00009126
InChI Key LYUUVYQGUMRKOV-UHFFFAOYSA-N
PubChem CID 76664
IUPAC Name diethyl 2,2-bis(prop-2-enyl)propanedioate
SMILES CCOC(=O)C(CC=C)(CC=C)C(=O)OCC

Specifications

Density 0.993
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.446
Quantity 50 g
Beilstein 1790529
Synonym diethyl diallylmalonate, diethyl 2,2-diallylmalonate, diallylmalonic acid diethyl ester, propanedioic acid, di-2-propenyl-, diethyl ester, 1,3-diethyl 2,2-bis prop-2-en-1-yl propanedioate, diethyl 2,2-diprop-2-enylpropane-1,3-dioate, diethyldiallylmalonate, malonic acid, diallyl-, diethyl ester, diethyl2,2-diallylmalonate, acmc-1cp69
Solubility Information Sparingly Soluble in water. (0.72g/L) (25°C)
Formula Weight 240.3
Percent Purity 98%
Chemical Name or Material Diethyl diallylmalonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.