missing translation for 'onlineSavingsMsg'
Learn More

Diethyl di-n-butylmalonate, 98+%, Thermo Scientific Chemicals

Product Code. 11312347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312347 25 g 25g
11302347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312347 Supplier Thermo Scientific Alfa Aesar Supplier No. L09902.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-75-8
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.385
MDL Number MFCD00026842
InChI Key WHKKUUPZLWUOIW-UHFFFAOYSA-N
Synonym diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
PubChem CID 69000
IUPAC Name diethyl 2,2-dibutylpropanedioate
SMILES CCCCC(CCCC)(C(=O)OCC)C(=O)OCC

Specifications

Density 0.95
Boiling Point 150°C (12 mmHg)
Refractive Index 1.433
Quantity 25 g
Beilstein 518421
Formula Weight 272.39
Percent Purity ≥98%
Chemical Name or Material Diethyl di-n-butylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.