missing translation for 'onlineSavingsMsg'
Learn More

(-)-Diethyl D-tartrate, 99%

Product Code. 11439126
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439126 5 g 5g
11449126 25 g 25g
11459126 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439126 Supplier Thermo Scientific Alfa Aesar Supplier No. A17992.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 13811-71-7 | C8H14O6 | 206.19 g/mol

Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl D(-)-tartrate is mainly used in chiral pharmaceuticals, chiral intermediates and chiral catalysts. It can be used with titanium in enolic asymmetric epoxidization. The compound d-alpha-tocopherol is the principal compound present in natural vitamin E sources. At present, d-alpha-tocopherol and other tocopherol derivatives are used in pharmaceuticals, foods and animal feeds. It is used as a food additive.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13811-71-7
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00064451
InChI Key YSAVZVORKRDODB-WDSKDSINSA-N
Synonym --diethyl d-tartrate, 2s,3s-diethyl 2,3-dihydroxysuccinate, diethyl d-tartrate, d---tartaric acid diethyl ester, diethyl d---tartrate, diethyl-d-tartrate, diethyl 2s,3s-2,3-dihydroxybutanedioate, 2s,3s--dihydroxybutane-1,4-dioic acid diethyl ester, --diethyl-d-tartrate, diethyl s-r*,r*-tartrate
PubChem CID 117410
IUPAC Name diethyl (2S,3S)-2,3-dihydroxybutanedioate
SMILES CCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC

Specifications

Melting Point 17°C
Density 1.205
Boiling Point 280°C
Flash Point 93°C (199°F)
Refractive Index 1.446
Quantity 5 g
Beilstein 1727143
Solubility Information Insoluble in water.
Optical Rotation −26.5° (c=1 in Water)
Formula Weight 206.19
Percent Purity 99%
Chemical Name or Material (-)-Diethyl D-tartrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.