missing translation for 'onlineSavingsMsg'
Learn More

Diethyl D-Malate, TRC

Product Code. 30388589
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388589 Supplier Toronto Research Chemicals Supplier No. TRCD4444151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl (2R)-2-hydroxybutanedioate
CAS 7554-28-1
Molecular Formula C8 H14 O5
Quantity 1 g
Synonym (2R)-2-Hydroxy-butanedioic Acid 1,4-Diethyl Ester, Hydroxy-Butanedioic Acid Diethyl Ester, (R)-Hydroxy-butanedioic Acid Diethyl Ester, Diethyl (2R)-Malate, Diethyl (R)-2-Hydroxysuccinate, Diethyl (R)-malate
InChI Formula InChI=1S/C8H14O5/c1-3-12-7(10)5-6(9)8(11)13-4-2/h6,9H,3-5H2,1-2H3/t6-/m1/s1
SMILES CCOC(=O)C[C@@H](O)C(=O)OCC
IUPAC Name diethyl (2R)-2-hydroxybutanedioate
Molecular Weight (g/mol) 190.194
Formula Weight 190.084
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.