missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyclohexane-1,1-dicarboxylate, 95%, Thermo Scientific™

Product Code. p-7032796
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15437357

Brand: Thermo Scientific Alfa Aesar H34412.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1139-13-5
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD02093457
InChI Key MLHUKQNAQSRVKI-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclohexanedicarboxylate, 1,1-cyclohexanedicarboxylic acid diethyl ester, 1,1-diethyl cyclohexane-1,1-dicarboxylate, cyclohexane-1,1-dicarboxylic acid diethyl ester, 1,1-cyclohexanedicarboxylic acid, diethyl ester, acmc-1c6ic, 1,1-bis ethoxycarbonyl cyclohexane, cyclohexanedicarboxylicaciddiethylester, diethyl 1,1-cyclohexanedicarboxylate #, 1,1-cyclohexanedicarboxylicacid, 1,1-diethyl ester
PubChem CID 249270
IUPAC Name diethyl cyclohexane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CCCCC1)C(=O)OCC

Specifications

Boiling Point 106°C to 108°C (1 mmHg)
Quantity 1 g
Formula Weight 228.29
Percent Purity 95%
Chemical Name or Material Diethyl cyclohexane-1,1-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.