missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Cyanophosphonate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Diethyl Cyanophosphonate |
| CAS | 2942-58-7 |
| Molecular Formula | C5H10NO3P |
| Quantity | 1 g |
| Synonym | Ethyl phosphorocyanidate, Cyanophosphonic Acid diethyl ester, DEPC, Diethoxyphosphoryl cyanide, Diethyl Cyanophosphate, Diethyl phosphorocyanidate, Diethylphosphoryl cyanide |
| InChI Formula | InChI=1S/C5H10NO3P/c1-3-8-10(7,5-6)9-4-2/h3-4H2,1-2H3 |
| SMILES | CCOP(=O)(OCC)C#N |
| Molecular Weight (g/mol) | 163.11 |
| Formula Weight | 163.0398 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction