missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanophosphonate, tech. 90%, Thermo Scientific™

Product Code. p-7046374
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11388975

Brand: Thermo Scientific Alfa Aesar L14107.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Diethyl cyanophosphonate is used for peptide synthesis. and for the phosphorylation of phenols. It is also used as an activating agent for fluorescent derivatization of carboxylic acids which allows separation by HPLC, to activate carboxylic acids for the C-acylation of active methylene compounds and as a reagent for the phosphorylation of phenols. It promotes the formation of amides and esters from amines or alcohols in the presence of, e.g. triethylamine, activates carboxylic acids towards nucleophiles

Solubility
Hydrolyzes in water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-58-7
Molecular Formula C5H10NO3P
Molecular Weight (g/mol) 163.11
MDL Number MFCD00010256
InChI Key ZWWWLCMDTZFSOO-UHFFFAOYSA-N
Synonym diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
PubChem CID 76257
IUPAC Name diethoxyphosphorylformonitrile
SMILES CCOP(=O)(OCC)C#N

Specifications

Density 1.075
Boiling Point 104°C to 105°C (19 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.401
Quantity 5 g
UN Number UN2922
Beilstein 1768938
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 163.11
Percent Purity 90%
Grade Technical
Chemical Name or Material Diethyl cyanophosphonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.