missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanophosphonate, tech. 90%, Thermo Scientific™

Product Code. 11388975
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388975 5 g 5g
11398975 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388975 Supplier Thermo Scientific Alfa Aesar Supplier No. L14107.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Diethyl cyanophosphonate is used for peptide synthesis. and for the phosphorylation of phenols. It is also used as an activating agent for fluorescent derivatization of carboxylic acids which allows separation by HPLC, to activate carboxylic acids for the C-acylation of active methylene compounds and as a reagent for the phosphorylation of phenols. It promotes the formation of amides and esters from amines or alcohols in the presence of, e.g. triethylamine, activates carboxylic acids towards nucleophiles

Solubility
Hydrolyzes in water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2942-58-7
Molecular Formula C5H10NO3P
Molecular Weight (g/mol) 163.11
MDL Number MFCD00010256
InChI Key ZWWWLCMDTZFSOO-UHFFFAOYSA-N
Synonym diethyl cyanophosphonate, diethyl phosphorocyanidate, diethylphosphorocyanidate, diethylcyanophosphate, diethyl cyanidophosphate, phosphorocyanidic acid, diethyl ester, diethoxy-phosphoryl cyanide, diethyl cyanophosphosphonate, cyanophosphonic acid diethyl ester, diethylcyanophosphonate
PubChem CID 76257
IUPAC Name diethoxyphosphorylformonitrile
SMILES CCOP(=O)(OCC)C#N

Specifications

Density 1.075
Boiling Point 104°C to 105°C (19 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.401
Quantity 5 g
UN Number UN2922
Beilstein 1768938
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 163.11
Percent Purity 90%
Grade Technical
Chemical Name or Material Diethyl cyanophosphonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.