missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Cyanomethylphosphonate, TRC

Product Code. 30385349
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385349 1 g 1g
30378725 5 g -
30380937 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385349 Supplier Toronto Research Chemicals Supplier No. TRCD4441651G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
Synonym P-(Cyanomethyl)phosphonic Acid Diethyl Ester, (Cyanomethyl)diethoxyphosphine Oxide, (Diethoxyphosphinyl)acetonitrile, Diethyl Cyanomethanephosphonate, O,O-Diethyl (Cyanomethyl)phosphonate, NSC 407826
IUPAC Name 2-diethoxyphosphorylacetonitrile
SMILES CCOP(=O)(CC#N)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H12NO3P/c1-3-9-11(8,6-5-7)10-4-2/h3-4,6H2,1-2H3
Formula Weight 177.0555
Chemical Name or Material Diethyl Cyanomethylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.