Learn More
Diethyl cyanomethylphosphonate, 96%
Description
Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.
Solubility
Miscible with water, chloroform, terahydrofuran, ethyl acetate and methylene chloride.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Density | 1.094 |
| Boiling Point | 132°C to 135°C (1.2 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.433 |
| Quantity | 100 g |
| UN Number | UN3278 |
| Beilstein | 1772981 |
| Solubility Information | Miscible with water,chloroform,terahydrofuran,ethyl acetate and methylene chloride. |
| Formula Weight | 177.14 |
| Percent Purity | ≥98% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.