missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Cromoglycate, TRC

Product Code. 30380796
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380796 250 mg
30385885 100 mg
30386407 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380796 Supplier Toronto Research Chemicals Supplier No. TRCD444130250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16150-45-1
Molecular Formula C27 H24 O11
Molecular Weight (g/mol) 524.47
Synonym Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate), Sodium Cromoglicate Imp. B (EP), Ethyl Cromoglicate, Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)bis(oxy)]bis(4-oxo-4H-chromene-2-carboxylate)
IUPAC Name ethyl 5-[3-(2-ethoxycarbonyl-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
SMILES CCOC(=O)C1=CC(=O)c2c(OCC(O)COc3cccc4OC(=CC(=O)c34)C(=O)OCC)cccc2O1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H24O11/c1-3-33-26(31)22-11-16(29)24-18(7-5-9-20(24)37-22)35-13-15(28)14-36-19-8-6-10-21-25(19)17(30)12-23(38-21)27(32)34-4-2/h5-12,15,28H,3-4,13-14H2,1-2H3
Formula Weight 524.1319
Chemical Name or Material Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.