missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chlorophosphate, TRC

Product Code. 30373868
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373868 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373868 Supplier Toronto Research Chemicals Supplier No. TRCD4440952.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 814-49-3
Molecular Formula C4H10ClO3P
Quantity 2.5 g
Synonym Phosphorochloridic Acid Diethyl Ester, Ethyl Phosphorochloridate, Chlorodiethoxyphosphine Oxide, Chlorodiethylphosphate, Diethoxyphosphorus Oxychloride, Diethyl Phosphoric Chloride, Ethyl Phosphorochloridate, O,O-Diethyl Chlorophosphate, Phosphoric Acid Diethyl Ester Chloride
InChI Formula InChI=1S/C4H10ClO3P/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3
SMILES CCOP(=O)(Cl)OCC
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
Molecular Weight (g/mol) 172.55
Formula Weight 172.0056
Chemical Name or Material Diethyl Chlorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.