missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chlorophosphate, 97+%

Product Code. 15446687
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446687 100 g 100g
15436687 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446687 Supplier Thermo Scientific Alfa Aesar Supplier No. H33820.22
  • ONLINE EXCLUSIVE PRICE
    £99.80
    £89.65 / 100g
    Save £10.15
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 814-49-3 | C4H10ClO3P | 172.545 g/mol

A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-49-3
Molecular Formula C4H10ClO3P
Molecular Weight (g/mol) 172.545
MDL Number MFCD00009075
InChI Key LGTLXDJOAJDFLR-UHFFFAOYSA-N
Synonym diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
PubChem CID 13139
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)Cl

Specifications

Density 1.194
Boiling Point 60°C (2 mmHg)
Flash Point 61°C (142°F)
Quantity 100 g
UN Number UN3278
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 172.55
Percent Purity ≥97%
Chemical Name or Material Diethyl chlorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.