missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (chloromethyl)phosphonate, 98%, Thermo Scientific Chemicals

Catalog Number 10193011
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
3167-63-3
C5H12ClO3P
186.57
MFCD00010189
MZBIWKMCTWJLPT-UHFFFAOYSA-N
diethyl chloromethyl phosphonate, diethyl chloromethylphosphonate, diethyl chloromethanephosphonate, phosphonic acid, chloromethyl-, diethyl ester, chloromethylphosphonic acid diethyl ester, phosphonic acid, p-chloromethyl-, diethyl ester, chloromethyl phosphonic acid diethyl ester, 1-chloromethyl ethoxy phosphoryl oxyethane, acmc-1cq3s
76633
1-[chloromethyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(CCl)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
3167-63-3
C5H12ClO3P
186.57
MFCD00010189
MZBIWKMCTWJLPT-UHFFFAOYSA-N
diethyl chloromethyl phosphonate, diethyl chloromethylphosphonate, diethyl chloromethanephosphonate, phosphonic acid, chloromethyl-, diethyl ester, chloromethylphosphonic acid diethyl ester, phosphonic acid, p-chloromethyl-, diethyl ester, chloromethyl phosphonic acid diethyl ester, 1-chloromethyl ethoxy phosphoryl oxyethane, acmc-1cq3s
76633
1-[chloromethyl(ethoxy)phosphoryl]oxyethane
CCOP(=O)(CCl)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2000g/mL
Boiling Point 109.0°C to 110.0°C (10.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Quantity 1 g
Specific Gravity 1.2
Formula Weight 186.57
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl (chloromethyl)phosphonate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl (chloromethyl)phosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'