missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl chloromalonate, 90%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 221.0°C to 222.0°C |
| Color | Colorless |
| Density | 1.2040g/mL |
| Flash Point | 113°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Linear Formula | C2H5O2CCH(Cl)CO2C2H5 |
| Refractive Index | 1.431 |
| Quantity | 25 mL |
| Specific Gravity | 1.204 |
| Show More |
Safety and Handling
Product Identifier
- Diethyl chloromalonate
- Danger
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction