missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Carbonate, TRC

Product Code. 30376818
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30376818

Brand: Toronto Research Chemicals TRCD44400050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Carbonate
CAS 105-58-8
Purity Grade Notes GC
Molecular Formula C5H10O3
Quantity 50 g
Synonym DEC, Diatol, Diatol (Carbonate), Diethyl Carbonate, Diethyl Ester Carbonic Acid, Ethyl Carbonate, Ethyl Carbonate ((EtO)2CO), Eufin, H-DEC, NSC 8849
InChI Formula InChI=1S/C5H10O3/c1-3-7-5(6)8-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)OCC
Molecular Weight (g/mol) 118.03
Formula Weight 118.063
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.