missing translation for 'onlineSavingsMsg'
Learn More

Diethyl carbonate, 99%, anhydrous, AcroSeal™

Product Code. 11925161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925161 100 mL 100mL
11915161 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925161 Supplier Thermo Scientific Acros Supplier No. 436131000
  • ONLINE EXCLUSIVE PRICE
    £45.20
    £37.65 / 100mL
    Save £7.55
    16% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-58-8 | C5H10O3 | 118.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-58-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00009107
InChI Key OIFBSDVPJOWBCH-UHFFFAOYSA-N
Synonym ethyl carbonate, carbonic acid, diethyl ester, carbonic acid diethyl ester, eufin, diatol, diaethylcarbonat, carbonic ether, ethoxyformic anhydride, diethylkarbonat, ethyl carbonate eto 2co
PubChem CID 7766
IUPAC Name diethyl carbonate
SMILES CCOC(=O)OCC

Specifications

Melting Point -43.0°C
Density 0.9750g/mL
Boiling Point 126.0°C to 128.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5O)2CO
Refractive Index 1.3830 to 1.3850
Quantity 100 mL
Merck Index 15,3835
Specific Gravity 0.975
Solubility Information Solubility in water: 18.8g/L.
Viscosity 0.827 mPa.s (20°C)
Formula Weight 118.13
Percent Purity 99%
Chemical Name or Material Diethyl carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl carbonate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.