missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Butylethylmalonate-d5, TRC

Product Code. 30388463
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388463 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30388463 Supplier Toronto Research Chemicals Supplier No. TRCD443877100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Butylethylmalonate-d5
CAS 145395-63-7
Molecular Formula C13H19D5O4
Quantity 100 mg
Synonym Butylethyl-d5-propanedioic Acid Diethyl Ester, 2-Butyl-2-ethylpropanedioic Acid 1,3-Diethyl Ester-d5, Butylethylmalonic Acid Diethyl Ester-d5, Butylethylpropanedioic Acid Diethyl Ester-d5, Diethyl Ethylbutylmalonate-d5, NSC 44910-d5, NSC 6791-d5
InChI Formula InChI=1S/C13H24O4/c1-5-9-10-13(6-2,11(14)16-7-3)12(15)17-8-4/h5-10H2,1-4H3/i2D3,6D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C(CCCC)(C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-butyl-2-(1,1,2,2,2-pentadeuterioethyl)propanedioate
Molecular Weight (g/mol) 249.36
Formula Weight 249.1988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.