missing translation for 'onlineSavingsMsg'
Learn More

Diethyl bromomalonate, 90+%

Product Code. 10734031 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10734031 100 mL Glass bottle 100mL
10510401 25 mL Glass bottle 25mL
10289843 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10734031 Supplier Thermo Scientific Acros Supplier No. 114191000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 685-87-0 | C7H11BrO4 | 239.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 685-87-0
Molecular Formula C7H11BrO4
Molecular Weight (g/mol) 239.07
MDL Number MFCD00009138
InChI Key FNJVDWXUKLTFFL-UHFFFAOYSA-N
Synonym diethyl bromomalonate, diethyl 2-bromomalonate, bromomalonic acid diethyl ester, ethyl bromomalonate, diethylbromomalonate, propanedioic acid, bromo-, diethyl ester, 1,3-diethyl 2-bromopropanedioate, diethyl bromopropanedioate, malonic acid, bromo-, diethyl ester, propanedioic acid, 2-bromo-, 1,3-diethyl ester
PubChem CID 69637
IUPAC Name diethyl 2-bromopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)Br

Specifications

Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 233°C to 235°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH(Br)CO2C2H5
Refractive Index 1.4520 to 1.456
Quantity 100 mL
Beilstein 02, 594
Specific Gravity 1.4
Solubility Information Solubility in water: immiscible
Formula Weight 239.07
Percent Purity 90+%
Physical Form Liquid
Chemical Name or Material Diethyl bromomalonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl bromomalonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.