missing translation for 'onlineSavingsMsg'
Learn More

Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog Number 11413384
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
1080-32-6
C11H17O3P
228.228
MFCD00009078
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
14122
diethoxyphosphorylmethylbenzene
CCOP(=O)(CC1=CC=CC=C1)OCC
This item is not returnable. View return policy
1080-32-6
C11H17O3P
228.228
MFCD00009078
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
14122
diethoxyphosphorylmethylbenzene
CCOP(=O)(CC1=CC=CC=C1)OCC

Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.094
Boiling Point 106°C to 108°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4975
Quantity 50 g
Beilstein 2580931
Solubility Information Insoluble in water.
Formula Weight 228.23
Percent Purity 99%
Chemical Name or Material Diethyl benzylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'