missing translation for 'onlineSavingsMsg'
Learn More

Diethyl aminomalonate hydrochloride, 98%

Product Code. 11424654
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424654 250 g 250g
11404654 10 g 10g
11414654 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424654 Supplier Thermo Scientific Alfa Aesar Supplier No. A13681.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13433-00-6 | C7H14ClNO4 | 211.642 g/mol

Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl aminomalonate hydrochloride is used as a pharmaceutical intermediate. Diethyl aminomalonate with formaldehyde generates an azomethine ylide which can undergo cycloadditions with 1,2-dipolarophiles to form pyrrolidines.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13433-00-6
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00012510
InChI Key GLFVNTDRBTZJIY-UHFFFAOYSA-N
Synonym diethyl aminomalonate hydrochloride, diethyl 2-aminomalonate hydrochloride, aminomalonic acid diethyl ester hydrochloride, 1,3-diethyl 2-aminopropanedioate hydrochloride, propanedioic acid, amino-, diethyl ester, hydrochloride, diethyl 2-aminopropanedioate hydrochloride, diethyl 2-aminomalonate hcl, diethyl aminomalonate hcl, diethyl 2-aminopropane-1,3-dioate, chloride, pubchem23977
PubChem CID 3084143
IUPAC Name diethyl 2-aminopropanedioate;hydrochloride
SMILES CCOC(=O)C(C(=O)OCC)N.Cl

Specifications

Melting Point ∼165°C (decomposition)
Assay Percent Range 98%
Quantity 250 g
Beilstein 3568037
Solubility Information Soluble in water.
Formula Weight 211.65
Percent Purity 98%
Chemical Name or Material Diethyl aminomalonate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.