missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allylmalonate, 96%

Product Code. 11307655
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307655 25 g 25g
11317655 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307655 Supplier Thermo Scientific Alfa Aesar Supplier No. L02286.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2049-80-1 | C10H16O4 | 200.234 g/mol

Diethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl allylmalonate is an important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals, agrochemicals and flavors and fragrances compounds.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-80-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00009155
InChI Key GDWAYKGILJJNBB-UHFFFAOYSA-N
Synonym diethyl allylmalonate, ethyl allylmalonate, diethyl 2-allylmalonate, allylmalonic acid diethyl ester, diethyl prop-2-enylpropanedioate, diethyl 2-prop-2-enyl malonate, malonic acid, allyl-, diethyl ester, propanedioic acid, 2-propenyl-, diethyl ester, diethyl 2-2-propenyl-1,3-propanedioate, allyl malonic acid
PubChem CID 74900
IUPAC Name diethyl 2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)C(=O)OCC

Specifications

Density 1.016
Boiling Point 222°C to 223°C
Flash Point 92°C (197°F)
Refractive Index 1.431
Quantity 25 g
Beilstein 1709466
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Diethyl allylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.