missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Adipate-d10, TRC

Product Code. 30383202
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383202 10 mg
30379632 50 mg
30370478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30383202 Supplier Toronto Research Chemicals Supplier No. TRCD45540710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 D10 H8 O4
Molecular Weight (g/mol) 212.309
Synonym Diethyl Adipate-d10, Diethyl Hexanedioate-d10, Ethyl Adipate-d10, Ethyl delta-Carboethoxyvalerate-d10, NSC 19160-d10, NSC 3363-d10
IUPAC Name bis(1,1,2,2,2-pentadeuterioethyl) hexanedioate
SMILES [2H]C([2H])([2H])C([2H])([2H])OC(=O)CCCCC(=O)OC([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3/i1D3,2D3,3D2,4D2
Formula Weight 212.183
Chemical Name or Material Diethyl Adipate-d10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.