missing translation for 'onlineSavingsMsg'
Learn More

Diethyl adipate, 99%

Product Code. 10625861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10625861 100 g Glass bottle 100g
10268383 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625861 Supplier Thermo Scientific Acros Supplier No. 407171000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-28-6 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-28-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009215
InChI Key VIZORQUEIQEFRT-UHFFFAOYSA-N
Synonym diethyl adipate, ethyl adipate, 1,6-diethyl hexanedioate, hexanedioic acid, diethyl ester, adipic acid, diethyl ester, adipic acid diethyl ester, hexanedioic acid, diethyl, diethylhexanedioic acid, diethylester kyseliny adipove, ethyl delta-carboethoxyvalerate
PubChem CID 8844
ChEBI CHEBI:34697
IUPAC Name diethyl hexanedioate
SMILES CCOC(=O)CCCCC(=O)OCC

Specifications

Melting Point -18.0°C
Color Colorless
Density 1.0000g/mL
Boiling Point 132.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2OCO(CH2)4COOCH3CH2
Refractive Index 1.4260 to 1.4280
Quantity 100 g
Beilstein 02, 652
Specific Gravity 1
Formula Weight 202.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl adipate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.