missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetonylsuccinate, 95%, Thermo Scientific™

Product Code. 15421819
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15421819

Brand: Thermo Scientific Alfa Aesar H64545.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187-74-2
Molecular Formula C11H18O5
Molecular Weight (g/mol) 230.26
MDL Number MFCD00071579
InChI Key AAPVOVKOHNCXJA-UHFFFAOYNA-N
Synonym diethyl 2-2-oxopropyl succinate, diethyl acetonylsuccinate, acetonylsuccinic acid diethyl ester, acetonyl succinic acid diethyl ester, butanedioic acid, 2-oxopropyl-, diethyl ester, 1,4-diethyl 2-2-oxopropyl butanedioate, acetonyl succinic, acetonylsuccinic acid, diethyl 2-acetonylbutanedioate, diethyl 2-2-oxopropyl succite
PubChem CID 5463816
SMILES CCOC(=O)CC(CC(C)=O)C(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.074 g/mL
Boiling Point 135°C (1.3 mmHg)
Refractive Index 1.441
Quantity 100 g
Beilstein 1911511
IUPAC Name diethyl 2-(2-oxopropyl)butanedioate
Formula Weight 230.26
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl acetonylsuccinate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.