missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetone-1,3-dicarboxylate, 96%

Product Code. 11404343
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404343 50 g 50g
11414343 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404343 Supplier Thermo Scientific Alfa Aesar Supplier No. A10933.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-50-0 | C9H14O5 | 202.21 g/mol

Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

Solubility
Insoluble in water and soluble in alcohol, ester, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Density 1.113
Boiling Point 134°C to 135°C (10 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.44
Quantity 50 g
Beilstein 640146
Solubility Information Insoluble in water and soluble in alcohol,ester,benzene.
Formula Weight 202.21
Percent Purity 96%
Chemical Name or Material Diethyl acetone-1,3-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.