missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetamidomalonate, 98+%

Product Code. 11417956
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417956 25 g 25g
11427956 100 g 100g
11437956 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417956 Supplier Thermo Scientific Alfa Aesar Supplier No. A14048.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1068-90-2 | C9H15NO5 | 217.221 g/mol

Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 2-Acetamidomalonate, is a versatile building block used for the synthesis of various pharmaceutical and biologically active compounds. It is an intermediate for the preparation of Novobiocin analogues as potential heat shock protein 90 inhibitors. It is also used as a important intermediates in syntheses of vitamins B1 and B6, barbiturates, non-steroidal anti-inflammatory agents, other numerous pharmaceuticals.

Solubility
Soluble in chloroform and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-90-2
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
MDL Number MFCD00009146
InChI Key ISOLMABRZPQKOV-UHFFFAOYSA-N
Synonym diethyl acetamidomalonate, diethyl 2-acetamidomalonate, diethyl acetaminomalonate, diethyl acetylaminomalonate, acetamidomalonic acid diethyl ester, 2-acetylaminomalonic acid diethyl ester, 1,3-diethyl 2-acetamidopropanedioate, diethyl acetylamino malonate, propanedioic acid, acetylamino-, diethyl ester, acetylamino propanedioic acid diethyl ester
PubChem CID 14041
IUPAC Name diethyl 2-acetamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC(=O)C

Specifications

Melting Point 96°C to 99°C
Boiling Point 185°C (20 mmHg)
Quantity 25 g
Beilstein 783883
Solubility Information Soluble in chloroform and methanol. Slightly soluble in water.
Formula Weight 217.22
Percent Purity ≥98%
Chemical Name or Material Diethyl acetamidomalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.