missing translation for 'onlineSavingsMsg'
Learn More

Diethyl a-Acetylglutarate, TRC

Product Code. 30383045
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383045 Supplier Toronto Research Chemicals Supplier No. TRCD4435105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl alpha-Acetylglutarate
CAS 1501-06-0
Molecular Formula C11H18O5
Quantity 5 g
Synonym 2-acetylglutaric Acid Diethyl ester, 2-acetylpentanedioic Acid Diethyl Ester, Diethyl 2-Acetylglutarate, Diethyl 2-Acetylpentanedioate, NSC 42535
InChI Formula InChI=1S/C11H18O5/c1-4-15-10(13)7-6-9(8(3)12)11(14)16-5-2/h9H,4-7H2,1-3H3
SMILES CCOC(=O)CCC(C(=O)C)C(=O)OCC
IUPAC Name diethyl 2-acetylpentanedioate
Molecular Weight (g/mol) 230.26
Formula Weight 230.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.