missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (6-methyl-2-pyridylaminomethylene)malonate, TRC

Product Code. 30383265
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383265 Supplier Toronto Research Chemicals Supplier No. TRCD444588100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl (6-methyl-2-pyridylaminomethylene)malonate
CAS 13250-95-8
Molecular Formula C7 H5 N O S
Quantity 100 mg
Synonym 2-[[(6-Methylpyridin-2-yl)amino]methylene]malonic Acid Diethyl Ester, Diethyl 2-[(6-Methyl-2-pyridyl)aminomethylene]malonate, Diethyl [[(6-Methyl-2-pyridyl)amino]methylene]malonate, NSC 169694
InChI Formula InChI=1S/C7H5NOS/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
SMILES O=C1Nc2ccccc2S1
IUPAC Name 3H-1,3-benzothiazol-2-one
Molecular Weight (g/mol) 151.186
Formula Weight 151.009
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.