missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5-Ethylpyridine-2,3-dicarboxylate, TRC

Product Code. 30385351
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385351 1 g -
30382459 25 g -
30381202 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385351 Supplier Toronto Research Chemicals Supplier No. TRCD4445601G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105151-39-1
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
Synonym 5-Ethyl-2,3-pyridinedicarboxylic Acid Diethyl Ester
SMILES CCOC(=O)c1cc(CC)cnc1C(=O)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H17NO4/c1-4-9-7-10(12(15)17-5-2)11(14-8-9)13(16)18-6-3/h7-8H,4-6H2,1-3H3
Formula Weight 251.1158
Chemical Name or Material Diethyl 5-Ethylpyridine-2,3-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.