missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate), Mikromol™

Product Code. 17963216
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17963216

Brand: Mikromol MM0039.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
CAS 16150-45-1
Analyte or Component Names Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate) (Ethyl Cromoglicate)
Shipping Condition Room Temperature
Molecular Formula C27 H24 O11
Certifications/Compliance ISO 17025
Synonym Sodium Cromoglicate Imp. B (EP), Cromolyn USP Related Compound B, Cromolyn USP RC B, Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)dioxy]bis(4-oxo-4H-1-benzopyran-2-carboxylate), Ethyl Cromoglicate, Diethyl 5,5'-[(2-hydroxypropane-1,3-diyl)bis(oxy)]bis(4-oxo-4H-chromene-2-carboxylate), Cromolyn Related Compound B, Sodium Cromoglicate Impurity B
InChI Formula InChI=1S/C27H24O11/c1-3-33-26(31)22-11-16(29)24-18(7-5-9-20(24)37-22)35-13-15(28)14-36-19-8-6-10-21-25(19)17(30)12-23(38-21)27(32)34-4-2/h5-12,15,28H,3-4,13-14H2,1-2H3
SMILES CCOC(=O)C1=CC(=O)c2c(OCC(O)COc3cccc4OC(=CC(=O)c34)C(=O)OCC)cccc2O1
IUPAC Name ethyl 5-[3-(2-ethoxycarbonyl-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Molecular Weight (g/mol) 524.47
Quantity 25 mg
Formula Weight 524.1319 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.