missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate, 96%, Thermo Scientific™

Product Code. 15428845
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15428845

Brand: Thermo Scientific Alfa Aesar H34228.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146381-87-5
Molecular Formula C13H16ClNO5S
Molecular Weight (g/mol) 333.78
MDL Number MFCD00414318
InChI Key AOFYGAIXNQKIHT-UHFFFAOYSA-N
Synonym diethyl 5-chloroacetyl amino-3-methylthiophene-2,4-dicarboxylate, 2,4-diethyl 5-2-chloroacetamido-3-methylthiophene-2,4-dicarboxylate, diethyl 5-2-chloroacetamido-3-methylthiophene-2,4-dicarboxylate, diethyl 5-chloroacetyl amino-3-methyl-2,4-thiophenedicarboxylate, diethyl 5-2-chloroacetyl amino-3-methyl-2,4-thiophenedicarboxylate, diethyl 5-2-chloroacetyl amino-3-methylthiophene-2,4-dicarboxylate, 2,4-thiophenedicarboxylicacid,5-2-chloroacetyl amino-3-methyl-,2,4-diethyl ester, 3-methyl-5-chloroacetyl amino thiophene-2,4-dicarboxylic acid diethyl ester, ethyl 2-2-chloroacetylamino-5-ethoxycarbonyl-4-methylthiophene-3-carboxyla te
PubChem CID 739580
IUPAC Name diethyl 5-[(2-chloroacetyl)amino]-3-methylthiophene-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(C)C(C(=O)OCC)=C(NC(=O)CCl)S1

Specifications

Melting Point 164°C to 169°C
Quantity 250 mg
Formula Weight 333.79
Percent Purity 96%
Chemical Name or Material Diethyl 5-(2-chloroacetamido)-3-methylthiophene-2,4-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.