missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-methoxybenzylphosphonate, 98+%

Product Code. 11311857
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311857 50 g 50g
11301857 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311857 Supplier Thermo Scientific Alfa Aesar Supplier No. L05226.18
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1145-93-3 | C12H19O4P | 258.254 g/mol

Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1145-93-3
Molecular Formula C12H19O4P
Molecular Weight (g/mol) 258.254
MDL Number MFCD00015662
InChI Key NKARVHNAACNYGE-UHFFFAOYSA-N
Synonym diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate
PubChem CID 4063826
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-methoxybenzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)OC)OCC

Specifications

Density 1.5
Boiling Point 136°C to 138°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5045
Quantity 50 g
Beilstein 2125148
Solubility Information Not miscible in water.
Formula Weight 258.25
Percent Purity ≥98%
Chemical Name or Material Diethyl 4-methoxybenzylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.