missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-chlorobenzylphosphonate, 98+%

Product Code. 11344394
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344394 10 g 10g
11354394 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344394 Supplier Thermo Scientific Alfa Aesar Supplier No. L00965.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39225-17-7 | C11H16ClO3P | 262.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39225-17-7
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
MDL Number MFCD00051568
InChI Key BVQPVBZRJSFOEZ-UHFFFAOYSA-N
Synonym diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
PubChem CID 99240
IUPAC Name 1-chloro-4-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC

Specifications

Density 1.082
Quantity 10 g
Refractive Index 1.5075
Beilstein 1346690
Boiling Point 153°C to 155°C (0.6 mmHg)
Formula Weight 262.67
Percent Purity ≥98%
Chemical Name or Material Diethyl 4-chlorobenzylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.