missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (4-chlorobenzyl)phosphonate, 98%, ACROS Organics™

Product Code. 10707212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10707212 5 g Glass bottle 5g
10717212 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707212 Supplier Thermo Scientific Acros Supplier No. 332260050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 39225-17-7
Molecular Formula C11H16ClO3P
Molecular Weight (g/mol) 262.67
MDL Number MFCD00051568
InChI Key BVQPVBZRJSFOEZ-UHFFFAOYSA-N
Synonym diethyl 4-chlorobenzylphosphonate, diethyl 4-chlorobenzyl phosphonate, diethyl 4-chlorophenyl methylphosphonate, 1-chloro-4-diethoxyphosphorylmethyl benzene, phosphonic acid, 4-chlorophenyl methyl-, diethyl ester, phosphonic acid, p-4-chlorophenyl methyl-, diethyl ester, acmc-1aifv, diethyl 4-chlorobenzylposphonate, diethyl 4-chlorobenzylphosphate
PubChem CID 99240
IUPAC Name 1-chloro-4-(diethoxyphosphorylmethyl)benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)Cl)OCC

Especificaciones

Color Colorless
Density 1.09g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5080 to 1.5100 (20°C, 589nm)
Assay Percent Range 98%
Formula Weight 262.67
Percent Purity ≥97.5%
Chemical Name or Material Diethyl (4-chlorobenzyl)phosphonate
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Diethyl (4-chlorobenzyl)phosphonate
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.