missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-bromophenylmalonate, Thermo Scientific™

Product Code. 11306136
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11306136 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306136 Supplier Thermo Scientific Maybridge Supplier No. MAY00148DA
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 93139-85-6
Packaging Amber Glass Bottle
Molecular Formula C13H15BrO4
Quantity 1 g
Synonym diethyl 4-bromophenylmalonate, diethyl 2-4-bromophenyl malonate, 1,3-diethyl 2-4-bromophenyl propanedioate, 2-4-bromophenyl malonic acid diethyl ester, diethyl 4-bromphenyl malonate, diethyl 4-bromophenyl malonate, 2-4-bromo-phenyl malonic acid diethyl ester, propanedioic acid, 4-bromophenyl-, diethyl ester
InChI Key NEEVEYWBDFTMKS-UHFFFAOYSA-N
SMILES CCOC(=O)C(C1=CC=C(C=C1)Br)C(=O)OCC
IUPAC Name diethyl 2-(4-bromophenyl)propanedioate
Molecular Weight (g/mol) 315.163
PubChem CID 10519382
Formula Weight 315.16
Percent Purity 95%
Chemical Name or Material Diethyl 4-bromophenylmalonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.