missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 4-aminobenzylphosphonate, 99%

Product Code. 11425946
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425946 25 g 25g
11415946 5 g 5g
11435946 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425946 Supplier Thermo Scientific Alfa Aesar Supplier No. B22279.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20074-79-7 | C11H18NO3P | 243.24 g/mol

Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20074-79-7
Molecular Formula C11H18NO3P
Molecular Weight (g/mol) 243.24
MDL Number MFCD00007913
InChI Key ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
Synonym diethyl 4-aminobenzylphosphonate, diethyl p-aminobenzylphosphonate, 4-diethoxyphosphorylmethyl aniline, p-aminobenzylphosphonic acid diethyl ester, diethyl p-aminobenzyl phosphonate, phosphonic acid, p-aminobenzyl-, diethyl ester, diethyl 4-aminophenyl methylphosphonate, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, timtec-bb sbb000830
PubChem CID 88358
SMILES CCOP(=O)(CC1=CC=C(N)C=C1)OCC

Specifications

Melting Point 91°C to 94°C
Quantity 25 g
Beilstein 2941613
Solubility Information Soluble in methylene chloride,acetone and benzene. Insoluble in water.
IUPAC Name 4-(diethoxyphosphorylmethyl)aniline
Formula Weight 243.24
Percent Purity 99%
Chemical Name or Material Diethyl 4-aminobenzylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.